Gene Transcripts

Gene Symbol
Ubqlnl
Entrez Gene ID
244179
Gene Name
ubiquilin-like
Gene Aliases
4922504M18Rik, Gm490
Chromosome Location
Chr. 7: 104148259 - 104150556 on Build GRCm38
UniGene ID
Mm.189668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198624.3 NP_941026.2 1 1196
GenBank mRNA AK076645.1 - 1 1187
BC113134.1 - 1 1127 40062178

View Full Product Details