Gene Transcripts
- Gene Symbol
- Fat2
- Entrez Gene ID
- 245827
- Gene Name
- FAT tumor suppressor homolog 2 (Drosophila)
- Gene Aliases
- EMI2, Fath2, Gm523, mKIAA0811
- Chromosome Location
- Chr. 11: 55250609 - 55336621 on Build GRCm38
- UniGene ID
- Mm.339363
- Species
- Mus musculus
View Full Product Details