Gene Transcripts

Gene Symbol
Rttn
Entrez Gene ID
246102
Gene Name
rotatin
Gene Aliases
4921538A15Rik, AI666264, C530033I08Rik, D230040K24
Chromosome Location
Chr. 18: 88971738 - 89131016 on Build GRCm38
UniGene ID
Mm.772
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175542.3 NP_780751.2 37 5260
GenBank mRNA AF265232.1 - 37 5260
AK032605.1 - 8 891
BC023916.1 - 7 782
BC049935.1 - 6 657

View Full Product Details