Gene Transcripts

Gene Symbol
Trpv3
Entrez Gene ID
246788
Gene Name
transient receptor potential cation channel, subfamily V, member 3
Gene Aliases
1110036I10Rik, AI644701, Nh, VRL3
Chromosome Location
Chr. 11: 73267489 - 73300368 on Build GRCm38
UniGene ID
Mm.347652
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145099.2 NP_659567.2 2 136
GenBank mRNA AF510316.1 - 2 136

View Full Product Details