Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153424.2 | NP_700473.2 | 14 | 2058 | |
GenBank mRNA | AF537131.1 | - | 12 | 1805 | |
AK038879.1 | - | 14 | 1813 | ||
AK084539.1 | - | 14 | 2060 | ||
AK172998.1 | - | 11 | 1371 | ||
AY118229.1 | - | 13 | 1750 | ||
BC138370.1 | - | 14 | 1783 | 8861390 |