Gene Transcripts

Gene Symbol
Angptl2
Entrez Gene ID
26360
Gene Name
angiopoietin-like 2
Gene Aliases
AI593246, AW260363, Arp2
Chromosome Location
Chr. 2: 33215951 - 33247725 on Build GRCm38
UniGene ID
Mm.208919
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011923.4 NP_036053.2 4 1612
GenBank mRNA AF125176.1 - 3 1099
AK037265.1 - 4 1505
AK155464.1 - 4 1613
AK187902.1 - 2 178
AK205240.1 - 2 178
AK214337.1 - 2 178
BC138609.1 - 4 1418 8861629
BC138610.1 - 4 1418 8861630

View Full Product Details