Gene Transcripts
- Gene Symbol
- Mapk10
- Entrez Gene ID
- 26414
- Gene Name
- mitogen-activated protein kinase 10
- Gene Aliases
- C230008H04Rik, JNK3, JNK3B1, JNK3B2, SAPK(beta), Serk2, p493F12, p54bSAPK
- Chromosome Location
- Chr. 5: 102907948 - 103211339 on Build GRCm38
- UniGene ID
- Mm.39253
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081567.1 |
NP_001075036.1
|
6, 7 |
984 |
|
NM_009158.2 |
NP_033184.2
|
6, 7 |
984 |
|
GenBank mRNA |
AB005665.1 |
- |
3, 4 |
367 |
|
AB096076.1 |
- |
4, 5 |
418 |
|
AB096077.1 |
- |
4, 5 |
418 |
|
AB096078.1 |
- |
4, 5 |
418 |
|
AB096079.1 |
- |
4, 5 |
418 |
|
AK036275.1 |
- |
6, 7 |
936 |
|
AK042368.1 |
- |
5, 6 |
549 |
|
AK082242.1 |
- |
6, 7 |
988 |
|
AK163206.1 |
- |
6, 7 |
935 |
|
AK163385.1 |
- |
3, 4 |
461 |
|
BC046625.1 |
- |
6, 7 |
923 |
6494163 |
L35236.1 |
- |
6, 7 |
727 |
|
View Full Product Details