Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253808.1 | NP_001240737.1 | 6 | 667 | |
NM_001253809.1 | NP_001240738.1 | 6 | 663 | ||
NM_012025.7 | NP_036155.1 | 7 | 798 | ||
GenBank mRNA | AB030252.1 | - | 6 | 647 | |
AF079974.1 | - | 6 | 608 | ||
AF212320.1 | - | 6 | 634 | ||
AF212321.1 | - | 7 | 765 | ||
AK053850.1 | - | 6 | 628 | ||
AK144608.1 | - | 7 | 766 | ||
AK144745.1 | - | 6 | 627 | ||
AK154929.1 | - | 7 | 786 | ||
AK162660.1 | - | 4 | 3867 | ||
AK165897.1 | - | 6 | 620 | ||
AK166084.1 | - | 7 | 756 | ||
AK166094.1 | - | 6 | 625 | ||
AK168020.1 | - | 6 | 624 | ||
AK168025.1 | - | 6 | 624 | ||
AK168170.1 | - | 6 | 624 | ||
AK168679.1 | - | 6 | 608 | ||
AK181912.1 | - | 3 | 192 | ||
AK183940.1 | - | 4 | 258 | ||
AK184356.1 | - | 2 | 97 | ||
AK193352.1 | - | 2 | 97 | ||
AK200143.1 | - | 2 | 97 | ||
AK207314.1 | - | 4 | 258 | ||
BC010715.1 | - | 6 | 613 | 3602242 |