Gene Transcripts

Gene Symbol
Nkain3
Entrez Gene ID
269513
Gene Name
Na+/K+ transporting ATPase interacting 3
Gene Aliases
C530048M05, E130310K16Rik
Chromosome Location
Chr. 4: 20118336 - 20780310 on Build GRCm38
UniGene ID
Mm.317473
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290410.1 NP_001277339.1 2 510
NM_172987.2 NP_766575.2 2 510
GenBank mRNA AK049776.1 - 2 710
AK053823.1 - 2 395
AK141456.1 - 2 410
BC075683.1 - 2 510 30613986
EF058052.1 - 2 157
EF058053.1 - 2 157
EF058054.1 - 2 157

View Full Product Details