Gene Transcripts
- Gene Symbol
- Rtcb
- Entrez Gene ID
- 28088
- Gene Name
- RNA 2',3'-cyclic phosphate and 5'-OH ligase
- Gene Aliases
- AI255213, AI463255, D10Wsu52e, FAAP, HSPC117
- Chromosome Location
- Chr. 10: 85938637 - 85957793 on Build GRCm38
- UniGene ID
- Mm.9257
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145422.4 |
NP_663397.1
|
9, 10 |
1275 |
|
GenBank mRNA |
AK145891.1 |
- |
9, 10 |
1272 |
|
AK145916.1 |
- |
9, 10 |
1272 |
|
AK149885.1 |
- |
9, 10 |
1230 |
|
AK150376.1 |
- |
9, 10 |
1234 |
|
AK150470.1 |
- |
9, 10 |
1234 |
|
AK151042.1 |
- |
9, 10 |
1268 |
|
AK151182.1 |
- |
9, 10 |
1234 |
|
AK151932.1 |
- |
9, 10 |
1234 |
|
AK151988.1 |
- |
9, 10 |
1268 |
|
AK152144.1 |
- |
9, 10 |
1234 |
|
AK153434.1 |
- |
9, 10 |
1234 |
|
AK167643.1 |
- |
9, 10 |
1261 |
|
AK168038.1 |
- |
9, 10 |
1269 |
|
AK172175.1 |
- |
9, 10 |
1269 |
|
AY424364.1 |
- |
9, 10 |
1268 |
|
BC003288.1 |
- |
9, 10 |
1277 |
3583081 |
View Full Product Details