Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023718.3 | NP_076207.1 | 10 | 1405 | |
GenBank mRNA | AF203701.1 | - | 10 | 1409 | |
AF213260.1 | - | 10 | 1409 | ||
AK142587.1 | - | 9 | 1263 | ||
BC071214.1 | - | 9 | 1255 | 5042082 |