Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013933.3 | NP_038961.2 | 1 | 320 | |
GenBank mRNA | AF115503.1 | - | 1 | 42 | |
AF157497.1 | - | 1 | 261 | ||
AK003576.1 | - | 1 | 261 | ||
AK012357.1 | - | 1 | 75 | ||
AK088132.1 | - | 1 | 299 | ||
AK159582.1 | - | 1 | 262 | ||
AK167381.1 | - | 1 | 321 | ||
AK168824.1 | - | 1 | 267 | ||
AK189268.1 | - | 49 | |||
BC003866.1 | - | 1 | 306 | 3490082 |