Gene Transcripts

Gene Symbol
Npsr1
Entrez Gene ID
319239
Gene Name
neuropeptide S receptor 1
Gene Aliases
9330128H10Rik, GPRA, Gpr154, MVTR, PGR14, VRR1
Chromosome Location
Chr. 9: 24097950 - 24317590 on Build GRCm38
UniGene ID
Mm.130824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175678.3 NP_783609.1 8 915
GenBank mRNA AK033957.1 - 8 916
AK041451.1 - 7 814
AK041501.1 - 2 211
AY299387.1 - 7 851

View Full Product Details