Gene Transcripts

Gene Symbol
Nwd1
Entrez Gene ID
319555
Gene Name
NACHT and WD repeat domain containing 1
Gene Aliases
A230063L24Rik
Chromosome Location
Chr. 8: 72646225 - 72716880 on Build GRCm38
UniGene ID
Mm.212927
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176940.5 NP_795914.3 7 2293
GenBank mRNA AK034868.1 - 1992
AK038792.1 - 6 1986
AK087622.1 - 7 2298
AK162888.1 - 7 2288
BC082552.1 - 7 2233
BC144786.1 - 5 1883 9053334
BC144787.1 - 5 1883
BC145703.1 - 5 1883 40131006

View Full Product Details