Gene Transcripts

Gene Symbol
Fndc7
Entrez Gene ID
320181
Gene Name
fibronectin type III domain containing 7
Gene Aliases
E230011A21Rik
Chromosome Location
Chr. 3: 108853678 - 108890008 on Build GRCm38
UniGene ID
Mm.244233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177091.5 NP_796065.2 5 1081
GenBank mRNA AK053989.1 - 5 944
AK054241.1 - 5 1081
AK165075.1 - 2 636
BC113188.1 - 5 887

View Full Product Details