Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029264.2 | NP_083540.1 | 14 | 1679 | |
GenBank mRNA | AK133079.1 | - | 14 | 1680 | |
AM690754.1 | - | 11 | 1563 | ||
BC042775.1 | - | 1 | 93 | ||
BC046539.1 | - | 1 | 93 | ||
BC137726.1 | - | 4 | 503 |