Gene Transcripts

Gene Symbol
Hcn4
Entrez Gene ID
330953
Gene Name
hyperpolarization-activated, cyclic nucleotide-gated K+ 4
Gene Aliases
Bcng3, Hcn3
Chromosome Location
Chr. 9: 58823512 - 58860955 on Build GRCm38
UniGene ID
Mm.151967
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081192.1 NP_001074661.1 1 742
GenBank mRNA AF064874.1 - 1 209
BC158106.1 - 1 842 9088182
BC158108.1 - 1 842 9088183

View Full Product Details