Gene Transcripts

Gene Symbol
Zfp36l3
Entrez Gene ID
333473
Gene Name
zinc finger protein 36, C3H type-like 3
Gene Aliases
AY661338
Chromosome Location
Chr. X: 53772686 - 53776394 on Build GRCm38
UniGene ID
Mm.327378
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009549.2 NP_001009549.1 1 2184
GenBank mRNA AY661338.1 - 1 2184

View Full Product Details