Gene Transcripts

Gene Symbol
Slc2a12
Entrez Gene ID
353169
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 12
Gene Aliases
GLUT-12, Glut12
Chromosome Location
Chr. 10: 22612723 - 22703880 on Build GRCm38
UniGene ID
Mm.473705
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178934.4 NP_849265.2 2 562
GenBank mRNA AJ549317.2 - 2 562
AK028970.1 - 2 535
AK031659.1 - 2 480
AK035972.1 - 2 443
AK143263.1 - 2 551
AY230881.1 - 2 461
BC096454.1 - 2 493 4223231
BC116314.1 - 2 423 40095492
BC116315.1 - 2 426 40095493
BC138316.1 - 2 409 8861336
BC138317.1 - 2 409 8861337

View Full Product Details