Gene Transcripts

Gene Symbol
Adam32
Entrez Gene ID
353188
Gene Name
a disintegrin and metallopeptidase domain 32
Gene Aliases
Chromosome Location
Chr. 8: 24836143 - 24948917 on Build GRCm38
UniGene ID
Mm.269223
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293693.1 NP_001280622.1 17 1869
NM_153397.2 NP_700446.2 17 1869
GenBank mRNA AF513715.1 - 17 1852
AK005759.1 - 17 1860
BC060983.1 - 17 1866 6772485

View Full Product Details