Gene Transcripts

Gene Symbol
Rc3h1
Entrez Gene ID
381305
Gene Name
RING CCCH (C3H) domains 1
Gene Aliases
5730557L09Rik, Gm551, N28103, mKIAA2025
Chromosome Location
Chr. 1: 160906411 - 160974976 on Build GRCm38
UniGene ID
Mm.329667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024952.2 NP_001020123.1 2 599
GenBank mRNA AK139648.1 - 2 571
AK173335.1 - 2 545
AY948287.1 - 2 540
BC138663.1 - 1 323 8861684

View Full Product Details