Gene Transcripts

Gene Symbol
Odf3l2
Entrez Gene ID
382384
Gene Name
outer dense fiber of sperm tails 3-like 2
Gene Aliases
Gm1157
Chromosome Location
Chr. 10: 79639526 - 79655710 on Build GRCm38
UniGene ID
Mm.244510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033473.2 NP_001028645.1 2 215
GenBank mRNA AK158063.1 - 2 216
BC147356.1 - 2 203 9056405
BC147357.1 - 2 203 9056398

View Full Product Details