Gene Transcripts

Gene Symbol
Specc1
Entrez Gene ID
432572
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1
Gene Aliases
2810012G08Rik, B230396K10Rik, Cytsb
Chromosome Location
Chr. 11: 61956763 - 62223013 on Build GRCm38
UniGene ID
Mm.458025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029936.3 NP_001025107.1 6 2353
NM_001281818.1 NP_001268747.1 8 2570
GenBank mRNA AK220465.1 - 6 2280

View Full Product Details