Gene Transcripts

Gene Symbol
Cenpo
Entrez Gene ID
52504
Gene Name
centromere protein O
Gene Aliases
2810429O05Rik, 8430427C03Rik, D12Ertd482e
Chromosome Location
Chr. 12: 4197058 - 4234294 on Build GRCm38
UniGene ID
Mm.70546
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134046.5 NP_598807.2 3 707
GenBank mRNA AK013198.1 - 3 379
AK018444.1 - 3 352
AK029226.1 - 3 708
AK134266.1 - 3 379
BC034399.1 - 3 366 4481047

View Full Product Details