Gene Transcripts

Gene Symbol
Parp8
Entrez Gene ID
52552
Gene Name
poly (ADP-ribose) polymerase family, member 8
Gene Aliases
2810430O08Rik, ARTD16, D13Ertd275e
Chromosome Location
Chr. 13: 116850842 - 117025913 on Build GRCm38
UniGene ID
Mm.274466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081009.1 NP_001074478.1 17, 18 1960
GenBank mRNA AK013206.1 - 1, 2 112
AK149889.1 - 16, 17 1927
AK149918.1 - 16, 17 1927
AK150208.1 - 16, 17 1927
AK150732.1 - 15, 16 1737
AK151510.1 - 15, 16 1738
AK152919.1 - 15, 16 1737
AK153169.1 - 15, 16 1737
AK153270.1 - 15, 16 1737
BC021315.1 - 7, 8 1094 5027417
BC021881.1 - 16, 17 1930 3710450
BC022679.1 - 2, 3 487
EF591877.1 - 1695

View Full Product Details