Gene Transcripts

Gene Symbol
Rwdd2b
Entrez Gene ID
53858
Gene Name
RWD domain containing 2B
Gene Aliases
ORF5
Chromosome Location
Chr. 16: 87433331 - 87440592 on Build GRCm38
UniGene ID
Mm.162406
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016924.2 NP_058620.2 3, 4 438
GenBank mRNA AF177771.1 - 3, 4 394
AK002746.1 - 3, 4 404
AK084966.1 - 3, 4 413
AK206976.1 - 1, 2 57
BC002143.1 - 3, 4 419 3484719

View Full Product Details