Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017397.3 | NP_059093.3 | 10, 11 | 1453 | |
GenBank mRNA | AF217000.1 | - | 10, 11 | 1328 | |
AF220454.1 | - | 10, 11 | 1456 | ||
AJ250027.1 | - | 10, 11 | 1328 | ||
AK154518.1 | - | 10, 11 | 1332 | ||
BC125471.1 | - | 10, 11 | 1453 | 40129986 | |
BC137721.1 | - | 10, 11 | 1453 | 8860740 |