Gene Transcripts

Gene Symbol
Lpar2
Entrez Gene ID
53978
Gene Name
lysophosphatidic acid receptor 2
Gene Aliases
Edg4, IPA2, LPA2
Chromosome Location
Chr. 8: 69822565 - 69831102 on Build GRCm38
UniGene ID
Mm.23253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020028.3 NP_064412.2 3 1189
GenBank mRNA AK051092.1 - 3 1139
AK144861.1 - 3 1190
BC060131.1 - 3 1325 6409729
BC064676.1 - 3 1325 6409729

View Full Product Details