Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019481.2 | NP_062354.2 | 12, 13 | 1378 | |
GenBank mRNA | AF199365.1 | - | 12, 13 | 1378 | |
AF199366.1 | - | 12, 13 | 1378 | ||
AK005153.1 | - | 6, 7 | 712 | ||
AK128899.1 | - | 12, 13 | 1378 | ||
BC022672.1 | - | 8, 9 | 757 | ||
BC040789.1 | - | 12, 13 | 1352 | ||
BC049981.1 | - | 12, 13 | 1360 | 5065026 |