Gene Transcripts

Gene Symbol
Uqcc1
Entrez Gene ID
56046
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 1
Gene Aliases
2310079L17Rik, 2410003P15Rik, 3110038N19Rik, Bfzb, Bfzp, Cbp3, Uqcc
Chromosome Location
Chr. 2: 155846886 - 155930343 on Build GRCm38
UniGene ID
Mm.158827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018888.4 NP_061376.2 10 1048
GenBank mRNA AF253516.1 - 10 795
AK010376.1 - 10 789
AK011152.1 - 10 802
AK011953.1 - 10 772
AK019425.1 - 6 386
AK077323.1 - 10 786
AK139488.1 - 10 1048
AK169413.1 - 10 785
BC050884.1 - 10 1001 6808756
BC057570.1 - 10 1001 6808756

View Full Product Details