Gene Transcripts

Gene Symbol
Dgke
Entrez Gene ID
56077
Gene Name
diacylglycerol kinase, epsilon
Gene Aliases
C87606, DAGK6, DGK
Chromosome Location
Chr. 11: 89037582 - 89060763 on Build GRCm38
UniGene ID
Mm.153695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019505.3 NP_062378.1 7, 8 1560
GenBank mRNA AF136744.1 - 7, 8 1197
AK044596.1 - 2296
AK049335.1 - 7, 8 1360
BC055867.1 - 7, 8 1492

View Full Product Details