Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019726.3 | NP_062700.2 | 6 | 753 | |
GenBank mRNA | AF153696.1 | - | 7 | 693 | |
AK185728.1 | - | 1 | 115 | ||
BC011317.1 | - | 6 | 753 | 3979292 | |
BC138879.1 | - | 6 | 709 | 8861901 | |
BC138880.1 | - | 6 | 710 | 8861902 |