Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019824.3 | NP_062798.1 | 7 | 550 | |
GenBank mRNA | AB038243.1 | - | 6 | 486 | |
AK003485.1 | - | 7 | 581 | ||
AK005437.1 | - | 7 | 569 | ||
BC013618.1 | - | 7 | 550 | 4013537 | |
BC054440.1 | - | 7 | 517 | 6395280 | |
BC055036.1 | - | 7 | 517 |