Gene Transcripts
- Gene Symbol
- Stx5a
- Entrez Gene ID
- 56389
- Gene Name
- syntaxin 5A
- Gene Aliases
- 0610031F24Rik, D19Ertd627e, Stx5
- Chromosome Location
- Chr. 19: 8741424 - 8755642 on Build GRCm38
- UniGene ID
- Mm.153061
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC021883.1 |
- |
2 |
722 |
|
View Full Product Details