Gene Transcripts

Gene Symbol
Diap3
Entrez Gene ID
56419
Gene Name
diaphanous homolog 3 (Drosophila)
Gene Aliases
4930417P13Rik, Dia2, Diaph3, Drf3, mDia2, mKIAA4117, p134MDia2
Chromosome Location
Chr. 14: 86655361 - 87141248 on Build GRCm38
UniGene ID
Mm.440585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019670.1 NP_062644.1 17 1974
GenBank mRNA AF094519.1 - 17 1974
AK137073.1 - 16 2063
AK138694.1 - 17 2004
AK161737.1 - 16 1992
AK166882.1 - 17 2030
AK220493.1 - 17 2038
BC003250.1 - 16 2008
BC028920.1 - 10 1275 4008236
BC085191.1 - 17 2084 30693873
BC086779.1 - 17 2049

View Full Product Details