Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020032.2 | NP_064416.1 | 7, 8 | 1266 | |
GenBank mRNA | AF176099.1 | - | 6, 7 | 1207 | |
AJ131889.1 | - | 7, 8 | 1266 | ||
AK003392.1 | - | 3, 4 | 402 | ||
AK133087.1 | - | 7, 8 | 1235 | ||
BC004767.1 | - | 6, 7 | 1216 | 3156259 |