Gene Transcripts
- Gene Symbol
- Psmd8
- Entrez Gene ID
- 57296
- Gene Name
- proteasome (prosome, macropain) 26S subunit, non-ATPase, 8
- Gene Aliases
- 6720456J22Rik, AA407360, AL033291, AL033322, AL033323, C76433
- Chromosome Location
- Chr. 7: 29174187 - 29180673 on Build GRCm38
- UniGene ID
- Mm.273152
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026545.3 |
NP_080821.3
|
2, 3 |
464 |
|
GenBank mRNA |
AK002921.1 |
- |
2, 3 |
303 |
|
AK003436.1 |
- |
2, 3 |
305 |
|
AK020132.1 |
- |
2, 3 |
323 |
|
AK027973.1 |
- |
2, 3 |
268 |
|
AK051257.1 |
- |
2, 3 |
316 |
|
AK150493.1 |
- |
2, 3 |
307 |
|
AK150982.1 |
- |
2, 3 |
271 |
|
AK153329.1 |
- |
2, 3 |
305 |
|
AK159216.1 |
- |
2, 3 |
273 |
|
AK159795.1 |
- |
2, 3 |
289 |
|
AK159926.1 |
- |
2, 3 |
299 |
|
AK166718.1 |
- |
2, 3 |
298 |
|
AK168886.1 |
- |
2, 3 |
272 |
|
BC004075.1 |
- |
2, 3 |
282 |
3591796 |
BC005717.1 |
- |
2, 3 |
192 |
2609781 |
CF732286.1 |
- |
2, 3 |
464 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK013113.1 |
- |
|
263 |
|
Chr. - - [-] |
View Full Product Details