Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022325.4 | NP_071720.1 | 4 | 723 | |
GenBank mRNA | AF136277.1 | - | 4 | 733 | |
AJ242663.1 | - | 4 | 733 | ||
AK002710.1 | - | 4 | 685 | ||
AK004095.1 | - | 4 | 721 | ||
AK008370.1 | - | 4 | 652 | ||
AK010912.1 | - | 4 | 719 | ||
AK150329.1 | - | 4 | 651 | ||
AK150894.1 | - | 4 | 658 | ||
AK150947.1 | - | 4 | 712 | ||
AK151440.1 | - | 4 | 662 | ||
AK151585.1 | - | 4 | 686 | ||
AK151994.1 | - | 4 | 696 | ||
AK153147.1 | - | 4 | 658 | ||
AK159118.1 | - | 4 | 669 | ||
AK159618.1 | - | 4 | 736 | ||
AK159826.1 | - | 4 | 658 | ||
AK160001.1 | - | 4 | 706 | ||
AK160028.1 | - | 4 | 706 | ||
AK160354.1 | - | 4 | 738 | ||
AK166047.1 | - | 4 | 707 | ||
AK170963.1 | - | 4 | 706 | ||
AK186484.1 | - | 1 | 33 | ||
BC008619.1 | - | 4 | 723 | 3588457 |