Gene Transcripts

Gene Symbol
Nisch
Entrez Gene ID
64652
Gene Name
nischarin
Gene Aliases
1200007D05Rik, 3202002H23Rik, AW494485, I-1
Chromosome Location
Chr. 14: 31170928 - 31206826 on Build GRCm38
UniGene ID
Mm.298728
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022656.2 NP_073147.2 10, 11 1350
GenBank mRNA AF315344.1 - 6, 7 785
AK004628.1 - 10, 11 1187
AK036043.1 - 10, 11 1205
AK049791.1 - 10, 11 1205
AK086880.1 - 10, 11 1102
AK160990.1 - 10, 11 1224
BC003270.1 - 10, 11 1212
BC066838.1 - 10, 11 1208
BC089329.1 - 5, 6 569
BC108364.1 - 5, 6 545
BC144980.1 - 6, 7 679

View Full Product Details