Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022992.2 | NP_075368.1 | 1 | 189 | |
GenBank mRNA | AF133912.1 | - | 1 | 185 | |
AF265214.1 | - | 1 | 160 | ||
AF421860.1 | - | 1 | 195 | ||
AK005259.1 | - | 1 | 186 | ||
AK008519.1 | - | 1 | 169 | ||
AK076519.1 | - | 1 | 186 | ||
AK077826.1 | - | 1 | 196 | ||
AK088205.1 | - | 1 | 184 | ||
BC003897.1 | - | 1 | 189 | 3600236 | |
D87211.1 | - | 1 | 232 |