Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164791.1 | NP_001158263.1 | 7 | 684 | |
NM_023057.5 | NP_075544.1 | 7 | 748 | ||
NM_178084.4 | NP_835185.2 | 7 | 684 | ||
GenBank mRNA | AB049731.1 | - | 6 | 531 | |
AB049732.1 | - | 6 | 531 | ||
AK045444.1 | - | 7 | 609 | ||
AK087363.1 | - | 7 | 624 | ||
AK132186.1 | - | 7 | 749 | ||
AK162819.1 | - | 7 | 549 | ||
BC023718.1 | - | 7 | 629 | 5346593 |