Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025610.3 | NP_079886.2 | 5, 6 | 683 | |
GenBank mRNA | AK006147.1 | - | 5, 6 | 719 | |
AK010381.1 | - | 5, 6 | 725 | ||
AK030209.1 | - | 5, 6 | 684 | ||
BC016106.1 | - | 3, 4 | 435 | 4921265 |