Gene Transcripts

Gene Symbol
Aggf1
Entrez Gene ID
66549
Gene Name
angiogenic factor with G patch and FHA domains 1
Gene Aliases
2010009L17Rik, 2310029P06Rik, AW112072, Peg3, VG5Q
Chromosome Location
Chr. 13: 95350683 - 95375357 on Build GRCm38
UniGene ID
Mm.36785
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025630.3 NP_079906.2 14 2283
GenBank mRNA AK009533.1 - 10 1246
AY500995.1 - 14 2256
BC027286.1 - 9 959
BC028451.1 - 1 418
BC052410.1 - 14 2260 5716440

View Full Product Details