Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023229.2 | NP_075718.2 | 4 | 779 | |
GenBank mRNA | AF168682.1 | - | 3 | 674 | |
AK002552.1 | - | 4 | 730 | ||
AK003940.1 | - | 1 | 22 | ||
AK038596.1 | - | 4 | 731 | ||
AK052667.1 | - | 4 | 724 | ||
AK139374.1 | - | 3 | 1497 | ||
AK151859.1 | - | 4 | 731 | ||
AK152369.1 | - | 4 | 780 | ||
AK159751.1 | - | 4 | 731 | ||
AK164317.1 | - | 737 | |||
AK166508.1 | - | 1319 | |||
AK167346.1 | - | 3 | 1491 | ||
BC013547.1 | - | 4 | 696 | 4218374 |