Gene Transcripts

Gene Symbol
Hspbap1
Entrez Gene ID
66667
Gene Name
Hspb associated protein 1
Gene Aliases
3830421G21Rik
Chromosome Location
Chr. 16: 35769663 - 35828462 on Build GRCm38
UniGene ID
Mm.45272
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175111.3 NP_780320.2 8 1242
GenBank mRNA AK017731.1 - 8 1197
AK028824.1 - 9 1303
AK076189.1 - 8 1209

View Full Product Details