Gene Transcripts

Gene Symbol
Asf1b
Entrez Gene ID
66929
Gene Name
anti-silencing function 1B histone chaperone
Gene Aliases
1700003K02Rik, AA409591
Chromosome Location
Chr. 8: 83955694 - 83970195 on Build GRCm38
UniGene ID
Mm.29680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024184.2 NP_077146.1 4 656
GenBank mRNA AK005646.1 - 4 689
AK019594.1 - 4 438
AK077005.1 - 4 682
AK077718.1 - 4 637
AK134906.1 - 4 844
AK150818.1 - 4 637
AK151951.1 - 4 637
AK165322.1 - 4 635
BC003428.1 - 4 656 2645378

View Full Product Details