Gene Transcripts
- Gene Symbol
- Lap3
- Entrez Gene ID
- 66988
- Gene Name
- leucine aminopeptidase 3
- Gene Aliases
- 2410015L10Rik, AA410100, LAP-3, Lap, Lapep, Pep-7, Pep-S, Pep7, Peps
- Chromosome Location
- Chr. 5: 45493374 - 45512691 on Build GRCm38
- UniGene ID
- Mm.286830
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024434.6 |
NP_077754.3
|
1, 2 |
184 |
|
GenBank mRNA |
AF334160.1 |
- |
1, 2 |
145 |
|
AK002819.1 |
- |
1, 2 |
120 |
|
AK005334.1 |
- |
1, 2 |
97 |
|
AK008600.1 |
- |
1, 2 |
71 |
|
AK010502.1 |
- |
1, 2 |
137 |
|
AK011604.1 |
- |
1, 2 |
70 |
|
AK089023.1 |
- |
1, 2 |
151 |
|
AK166958.1 |
- |
1, 2 |
68 |
|
AK169032.1 |
- |
1, 2 |
70 |
|
BC016536.1 |
- |
1, 2 |
184 |
3488469 |
CT010237.1 |
- |
1, 2 |
98 |
|
CT010301.1 |
- |
1, 2 |
98 |
|
View Full Product Details