Gene Transcripts

Gene Symbol
Tbc1d2b
Entrez Gene ID
67016
Gene Name
TBC1 domain family, member 2B
Gene Aliases
1810061M12Rik, AI480956, AV021536, AV023399, AW491493, mKIAA1055
Chromosome Location
Chr. 9: 90202047 - 90270769 on Build GRCm38
UniGene ID
Mm.335347
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194334.2 NP_919315.2 4, 5 905
GenBank mRNA AK089175.1 - 4, 5 871
AK129273.1 - 4, 5 891
AK156788.1 - 4, 5 892

View Full Product Details