Gene Transcripts

Gene Symbol
Gatm
Entrez Gene ID
67092
Gene Name
glycine amidinotransferase (L-arginine:glycine amidinotransferase)
Gene Aliases
1810003P21Rik, AI314789, AT
Chromosome Location
Chr. 2: 122594473 - 122611277 on Build GRCm38
UniGene ID
Mm.29975
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025961.5 NP_080237.1 8 1135
GenBank mRNA AK007325.1 - 8 1135
AK145862.1 - 8 1137
AK145914.1 - 8 1137
AK146052.1 - 8 1134
AK146111.1 - 8 1136
AK151313.1 - 8 1134
AK152069.1 - 8 1136
AK153450.1 - 8 1120
BC003879.1 - 8 1140 3495871

View Full Product Details