Gene Transcripts

Gene Symbol
Tmem19
Entrez Gene ID
67226
Gene Name
transmembrane protein 19
Gene Aliases
2810428F02Rik, AW125171
Chromosome Location
Chr. 10: 115340739 - 115362524 on Build GRCm38
UniGene ID
Mm.29647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133683.3 NP_598444.1 3, 4 486
GenBank mRNA AK018383.1 - 3, 4 435
AK034298.1 - 3, 4 492
AK036228.1 - 2, 3 563
AK042426.1 - 2, 3 541
AK079735.1 - 344
AK136778.1 - 475
AK138931.1 - 3, 4 474
AK155275.1 - 3, 4 447
AK170143.1 - 3, 4 446
AK171087.1 - 3, 4 446
AK213890.1 - 2, 3 232
BC016895.1 - 3, 4 488 4239007

View Full Product Details